Artesunate
Supplier: Thermo Fisher Scientific
Warning
460340050 460340250
ACRO460340050EA 39.5 EUR
ACRO460340050 ACRO460340250
Artesunate
Artesunate
Formula: C₁₉H₂₈O₈
Melting Pt: 131…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866204
CAS Number: 88495-63-0
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %