2-Bromobornane, Technical Grade
Supplier: Thermo Fisher Scientific
460740050
460740250
ACRO460740050EA
50.9
EUR
ACRO460740050
ACRO460740250
2-Bromobornane, Technical Grade
2-Bromobornane
Formula:
C₁₀H₁₇Br Boiling Pt: 94…96 °C (15 mmHg) Storage Temperature: Refrigerator |
MDL Number:
MFCD00066594 CAS Number: 4443-48-5 |
Specification Test Results
Appearance | Off-white to brown Low melting solid |
Infrared spectrum | Conforms |
GC | ≥78.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...