1,8-Diazafluoren-9-one
Supplier: Thermo Fisher Scientific
Warning
461310010 461310050
ACRO461310010EA 338 EUR
ACRO461310010 ACRO461310050
1,8-Diazafluoren-9-one
1,8-Diazafluoren-9-one
Formula: C₁₁H₆N₂O
Melting Pt: 229…233 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082335
CAS Number: 54078-29-4
Order Now
Specification Test Results
Appearance Yellow to orange Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
Dye content ≥70.0 %