Universal dust mask, Cyrano®
Supplier: DELTALYOVALMY
CYRANO-XC CYRANO-XI12
VALMCYRANO-XCEA 78.6 EUR
VALMCYRANO-XC VALMCYRANO-XI12
Universal dust mask, Cyrano®
Masks Hygiene Masks
The FFP3 cup-shaped mask is both reassuring and light weight. The respiratory tract is shrouded in 10 g of comfort.
  • Exceptional breathing comfort, equivalent to an FFP1 mask
  • Airproof lining at the nose making it completely airtight from the very first inhalation
  • No need to ajust a nose piece
  • Perfect seal is guaranteed during the whole working time
  • 100% recyclable
Inhalation of 95 l/min
Certifications: EN149:2001 + A1:2009, EN14683:2006, EFB 99,9%
Packaging: Dispenser box of 10 or blister of 5.