pH electrodes, InLab® ISM
Supplier: Mettler - Toledo
51344102 51344103 51344104 51344055 51344155 51344211 51344072 51344172 51343156 30014096 30247850 30248830 30248832
662-1153EA 493 EUR
662-1153 662-1750 662-1910 662-1234 662-1236 662-1239 662-1235 662-1238 662-1245 663-0198 662-2032 662-2037 662-2038
pH electrodes, InLab® ISM
Electrodes pH Electrodes
High performance electrodes combining robustness with state-of-the-art measurement technology. Fully traceable.
  • Calibration data and Sensor ID are automatically transferred to the meter
  • New calibration data are stored in the sensor
  • Back-up certificate guaranteed: Initial factory calibration is stored in the sensor
  • Conclusive calibration history: The last five calibrations are stored in the sensor
  • Easy lifetime monitoring: The maximum temperature that the sensor has been exposed to is monitored automatically
The revolutionary Intelligent Sensor Management system (ISM) offers the user an even greater level of security. Calibration data is stored on the chip in the sensor and is used automatically when the sensor is connected to the meter. This ensures that the meter always uses the latest calibration data.
MultiPin™: pH / temperature
With integrated NTC 30 kOhm temperature probe (except InLab® Ultra-Micro-ISM).
* Without NTC 30 kOhm temperature probe.
Ordering information: Supplied with test certificate.