Anti-BAK Rabbit Polyclonal Antibody
Supplier: ENZO LIFE SCIENCES
ADIAAP030D ADIAAP030F
ENZOADIAAP030DEA 290 EUR
ENZOADIAAP030D ENZOADIAAP030F
Anti-BAK Rabbit Polyclonal Antibody
Antibodies
Bax is an appromimately 22 kDa pro-apoptotic protein belonging to the Bcl-2 family of proteins, which are characterized by the presence of at least one Bcl-2 homology (BH) domains. Bax normally resides in the cytoplasm, but following an apoptotic stimuli, Bax undergoes a conformational change and translocates to mitochondrial membranes where it inserts and mediates the release of cytochrome c. This activates caspase-3 and perpetuates the apoptotic cascade.
  • For Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human, hamster, monkey and pig Bak. Detects a band of ~30kDa by Western blot.
Type: Primary
Antigen: BAK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human