Anti-Myosin Mouse Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
ALXBC1040SL001 ALXBC1040SL005
ENZOALXBC1040SL001EA 256 EUR
ENZOALXBC1040SL001 ENZOALXBC1040SL005
Anti-Myosin Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2a

  • For Immunohistochemistry, Western Blot

Recognizes the S1/S2 junction of human ventricular myosin heavy chain beta.

Type: Primary
Antigen: MYOSIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR