Anti-PI31 Rabbit Polyclonal Antibody
Supplier: ENZO LIFE SCIENCES
BMLPW97100025 BMLPW97100100
ENZOBMLPW97100025EA 131 EUR
ENZOBMLPW97100025 ENZOBMLPW97100100
Anti-PI31 Rabbit Polyclonal Antibody
Antibodies
Recombinant PI31 has been shown to compete with the proteasome regulators PA28 and 19S PA700 for binding to 20S, indicating that PI31 binds the 20S α-rings and may function by hindering substrate access to the 20S catalytic channel.
  • For Western Blot
Recognizes human and mouse PI31.
Type: Primary
Antigen: PI31
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human