Anti-APP Mouse Monoclonal Antibody [clone: LN27]

Supplier: ENZO LIFE SCIENCES
ENZABS1260200 ENZABS1260500 ENZABS1261000
ENZOENZABS1260200EA 828 EUR
ENZOENZABS1260200 ENZOENZABS1260500 ENZOENZABS1261000
Anti-APP Mouse Monoclonal Antibody [clone: LN27]
Antibodies
Recommended Applications: IHC, WB

Species reactivity: Human

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: LN27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR