ImmunoSet® PDI ELISA development set
Supplier: Enzo Life Sciences
ImmunoSet®
ALEXADI-960-072EA 971 EUR
ALEXADI-960-072 ENZOADI960072
ImmunoSet® PDI ELISA development set
Assays ELISAs
For the quantitative determination of PDI in cell lysates, culture supernatants, plasma, and tissue of human, mouse, and rat origin.
The ImmunoSet® PDI ELISA development set contains sufficient reagents for the development of 5×96 well ELISA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own ELISA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules.
Ordering information: Contains Capture antibody, Standard, Detection antibody, SA-HRP.