Histaminergic ligand library, Screen-Well®
Supplier: ENZO LIFE SCIENCES
BML28160100 BML28160500
ENZOBML28160100EA 545 EUR
ENZOBML28160100 ENZOBML28160500
Histaminergic ligand library, Screen-Well®
Cell Culture Supplements and Additives
The Screen-Well® histaminergic and melatonin ligands plate contains 41 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Histaminergic and Melatonin Ligands plate is 1 plate of the larger neurotransmitter library which contains over 680 ligands.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
Species
Source
Learn more