Wnt pathway library, Screen-Well®
Supplier: ENZO LIFE SCIENCES
BML28380100 BML28380500
ENZOBML28380100EA 1700 EUR
ENZOBML28380100 ENZOBML28380500
Wnt pathway library, Screen-Well®
Cell Culture Supplements and Additives
The Screen-Well® Wnt pathway library v 2.0 is a focused collection of 71 compounds with defined and diverse Wnt pathway activity, including activators and inhibitors of Wnts, Dishevelled, GSK-3β and other Wnt pathway kinases, TCF/β-catenin, DKK, LRP, Axin, Porcupine, and more. Compounds are dissolved in DMSO at 10 mM or 1 mM and aliquoted into deep-well plates at 100 or 500 µl per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-Wnt pathway molecules in cells as well as for use in in vitro applications.
The Screen-Well® Wnt Pathway Library v 2.0 is a focused collection of 71 compounds with defined and diverse Wnt pathway activity, including activators and inhibitors of Wnts, Dishevelled, GSK-3β and other Wnt pathway kinases, TCF/β-catenin, DKK, LRP, Axin, Porcupine, and more. Compounds are dissolved in DMSO at 10mM or 1mM and aliquoted into deep-well plates at 100 or 500µL per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-Wnt pathway molecules in cells as well as for use in in vitro applications.
Ordering information: Contains 71 compounds with defined and diverse Wnt pathway activity. A variety of structurally and mechanistically different compound classes are included.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
Concentration DMSO solutions (10 mM ) except for Bafilomycin A which is provided at 1 mM in DMSO
Species
Source
Learn more