Sliding shelf racks for cryoboxes, Comfort
Supplier: TENAK
TE24312 TE24310 TE24304 TE24302 TE24294 TE24288 TE24286 TE24284 TE24278 TE24272 TE24276 TE24234 TE24266 TE24264 TE24262 TE24256 TE24254 TE24252 TE24250 TE24236 TE24274 TE24232 TE24230 TE24224 TE24222 TE24220 TE24238 TE24334 TE24122 TE24120 TE24124 TE24126 TE24130 TE24132 TE24134 TE24136 TE24138 TE24140 TE24150 TE24152 TE24154 TE24156 TE24296 TE24320 TE24322 TE24330 TE24332 TE24340 TE24342 TE24232B TE24234B TE24236B TE24220B TE24222B TE24224B TE24230B TE24238B TE24250B TE24252B TE24254B TE24256B TE24262B TE24264B TE24266B TE24272B TE24274B TE24276B TE24278B TE24284B TE24286B TE24288B TE24294B TE24296B TE24302B TE24304B TE24310B TE24312B TE24254-S
479-0784EA 218 EUR
479-0784 479-0783 479-0782 479-0781 479-0780 479-0779 479-0778 479-0777 479-0776 479-0773 479-0775 479-0763 479-0772 479-0771 479-0770 479-0769 479-0768 479-0767 479-0766 479-0764 479-0774 479-0762 479-0761 479-0760 479-0759 479-0758 479-0765 479-1677 479-1759 479-1377 479-1760 479-1761 479-1762 479-1763 479-1764 479-1765 479-1766 479-1767 479-1644 479-1645 479-1646 479-1647 479-1321 479-1673 479-1674 479-1675 479-1676 479-1678 479-1679 479-1006 479-1651 479-1652 479-1648 479-1005 479-1649 479-1650 479-1653 479-1654 479-1007 479-1655 479-1656 479-1657 479-1658 479-1659 479-1660 479-1661 479-1662 479-1663 479-1664 479-1665 479-1666 479-1667 479-1668 479-1669 479-1670 479-1671 479-1672 479-1839
Sliding shelf racks for cryoboxes, Comfort
Racks Cryogenic Box Racks
Stainless steel.
  • Racks for five different box heights are available
  • Can be used with all freezer brands
  • Fits all standard cryoboxes
For upright freezers. These racks ensure a more ergonomic operating process. The cryoboxes are placed in drawers in the sliding shelf rack, increasing flexibility. Measure the inside height and depth of your freezer in order to choose the right rack.
Ordering information: Delivered with or without cryoboxes.