Polydimethylsiloxane, 50 cSt, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dimethylpolysiloxane, PDMS, Silicone oil, Dimethicone
M-Clarity™ quality level = MQ200
378356-1L 378356-250ML
SIAL378356-1LEA 399 EUR
SIAL378356-1L SIAL378356-250ML
Polydimethylsiloxane, 50 cSt, Sigma-Aldrich®
Polydimethylsiloxane
Formula: (CH₃)₃SiO(Si(CH₃)₂O)n
Boiling Pt: >140 °C (0.002 mmHg)) (lit.)
Density: 0.96 g/ml
MDL Number: MFCD00084411
CAS Number: 63148-62-9