Polydimethylsiloxane, 350 cSt, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dimethylpolysiloxane, PDMS, Silicone oil, Dimethicone
M-Clarity™ quality level = MQ100
378372-1L 378372-250ML
SIAL378372-1LEA 409 EUR
SIAL378372-1L SIAL378372-250ML
Polydimethylsiloxane, 350 cSt, Sigma-Aldrich®
Polydimethylsiloxane
Formula: (CH₃)₃SiO(Si(CH₃)₂O)n
Boiling Pt: >140 °C (0.002 mmHg)) (lit.)
Density: 0.968 g/ml (25 °C)
MDL Number: MFCD00084411
CAS Number: 63148-62-9