Restriction enzymes, NotI, 10 U/µl, Fermentas
Supplier: Thermo Fisher Scientific
ER0591 ER0592 ER0595
ER0591EA 73.5 EUR
ER0591 ER0592 ER0595
Restriction enzymes, NotI, 10 U/µl, Fermentas
Nucleic Acid Reagents Restriction Enzymes
NotI restriction enzyme recognises GC^GGCCGC sites and cuts best at 37 °C in O buffer.
  • Superior quality, stringent quality control and industry leading manufacturing process
  • Convenient colour-coded five buffer system
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Applications: Molecular cloning, restriction site mapping, genotyping, southern blotting, restriction fragment length polymorphism (RFLP), SNP.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Enzyme name NotI
Source
Overhang type 5' overhang
Compatible buffer FastDigest Green Buffer or FastDigest Buffer (colorless)