Deuterium, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

361860-10L 361860-25L 361860-45L
SIAL361860-10LEA 194 EUR
SIAL361860-10L SIAL361860-25L SIAL361860-45L
Deuterium, Sigma-Aldrich®
Deuterium
Formula: D₂
MW: 4 g/mol
Boiling Pt: –249.6 °C (1013 hPa)
Melting Pt: –254.43 °C
Density: 0.169 g/cm³ (–253 °C)
Flash Pt: –240 °C
Storage Temperature: Ambient
CAS Number: 7782-39-0
EINECS: 231-952-7
UN: 1957
ADR: 2
Merck Index: 12,02983

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR