Cesium, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

239240-1G 239240-5G
SIAL239240-1GEA 139 EUR
SIAL239240-1G SIAL239240-5G
Cesium, Sigma-Aldrich®
Cesium
Formula: Cs
Boiling Pt: 705 °C (1013 hPa)
Melting Pt: 28.5 °C
Density: 1.873 g/cm³ (20 °C)
MDL Number: MFCD00134037
CAS Number: 7440-46-2
EINECS: 231-155-4
UN: 1407
ADR: 4.3,I
Merck Index: 12,02051

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR