Lithium, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
320080-100G 320080-25G
SIAL320080-100GEA 480 EUR
SIAL320080-100G SIAL320080-25G
Lithium, Sigma-Aldrich®
Lithium
Formula: Li
MW: 6.94 g/mol
Boiling Pt: 1317
Melting Pt: 179
Density: 0.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134051
CAS Number: 7439-93-2
EINECS: 231-102-5
UN: 1415
ADR: 4.3 I
Merck Index: 13,05542