Lithium, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
444456-10G 444456-50G
SIAL444456-10GEA 56.7 EUR
SIAL444456-10G SIAL444456-50G
Lithium, Sigma-Aldrich®
Lithium
Formula: Li
Boiling Pt: 1317 °C (1013 hPa)
Melting Pt: 179 °C
Density: 0.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134051
CAS Number: 7439-93-2
EINECS: 231-102-5
UN: 1415
ADR: 4.3,I
Merck Index: 13,05542