Lithium, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

499811-100G 499811-25G
SIAL499811-100GEA 270 EUR
SIAL499811-100G SIAL499811-25G
Lithium, Sigma-Aldrich®
Lithium
Formula: Li
MW: 6.94 g/mol
Boiling Pt: 1317
Melting Pt: 179
Density: 0.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134051
CAS Number: 7439-93-2
EINECS: 231-102-5
UN: 1415
ADR: 4.3 I
Merck Index: 13,05542

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR