Calcium, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

441872-25G 441872-5G
SIAL441872-25GEA 1260 EUR
SIAL441872-25G SIAL441872-5G
Calcium, Sigma-Aldrich®
Calcium
Formula: Ca
MW: 40.08 g/mol
Boiling Pt: 1484
Melting Pt: 850
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085314
CAS Number: 7440-70-2
EINECS: 231-179-5
UN: 1401
ADR: 4.3 II
Merck Index: 13,01644

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR