Magnesium ≥99.8% (metals basis), granules -12+50 mesh
Supplier: Thermo Fisher Scientific
Danger
00870.36 00870.A1 00870.A3
00870.36EA 124 EUR
00870.36 00870.A1 00870.A3
Magnesium ≥99.8% (metals basis), granules -12+50 mesh
Magnesium
Purity: 99.8% CAS No.: 7439-95-4 Appearance: Silver-grey Odour: Odourless UN No.: 1869 MDL No.: MFCD00085308
Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1107
Melting Pt: 651
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1 III
Merck Index: 13,05675