Magnesium ≥99.95% (metals basis), slug, annealed, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Fisher Scientific
43297.09 43297.18
43297.09EA 149 EUR
43297.09 43297.18
Magnesium ≥99.95% (metals basis), slug, annealed, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Magnesium
Purity: 99.95% CAS No.: 7439-95-4 Physical Form: ≈0.09g/slug Odour: Odourless MDL No.: MFCD00085308
Formula: Mg
Boiling Pt: 1107 °C (1013 hPa)
Melting Pt: 651 °C
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III
Merck Index: 13,05675