Magnesium ≥99.9% (metals basis), rods, Length: 25 mm (1 in), Ø 7.9 mm (0.31 in)
Supplier: Thermo Fisher Scientific
43355.KF 43355.KM 43355.KJ
43355.KFEA 44.6 EUR
43355.KF 43355.KM 43355.KJ
Magnesium ≥99.9% (metals basis), rods, Length: 25 mm (1 in), Ø 7.9 mm (0.31 in)
Magnesium
Purity: 99.9% CAS No.: 7439-95-4 Physical Form: ≈2.1g/piece Odour: Odourless MDL No.: MFCD00085308
Formula: Mg
Boiling Pt: 1107 °C (1013 hPa)
Melting Pt: 651 °C
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III
Merck Index: 13,05675