Magnesium, granules, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
254126-2KG 254126-500G
SIAL254126-2KGEA 402 EUR
SIAL254126-2KG SIAL254126-500G
Magnesium, granules, Sigma-Aldrich®
Magnesium
Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1107
Melting Pt: 651
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1 III
Merck Index: 13,05675