Magnesium, granules, Sigma-Aldrich®
Supplier: Merck
63040-1KG-F
63040-250G-F
SIAL63040-1KG-FEA
233
EUR
SIAL63040-1KG-F
SIAL63040-250G-F
Magnesium, granules, Sigma-Aldrich®
Magnesium
Formula:
Mg MW: 24.3 g/mol Boiling Pt: 1107 Melting Pt: 651 Density: 1.74 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1869 ADR: 4.1 III Merck Index: 13,05675 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...