Magnesium ≥99%, turnings
Supplier: Thermo Fisher Scientific
Danger
L08120.22 L08120.36 L08120.0E
L08120.22EA 25.7 EUR
L08120.22 L08120.36 L08120.0E
Magnesium ≥99%, turnings
Magnesium
Purity: 99+% CAS No.: 7439-95-4 Odour: Odourless UN No.: 1869 MDL No.: MFCD00085308
Formula: Mg
Boiling Pt: 1091 °C
Melting Pt: 650 °C
Density: 1.738
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III