Aluminium ≥99.9% (metals basis), granules 8-12 mm (0.32-0.47 in)
Supplier: Thermo Fisher Scientific
Synonyms: Aluminum
10093.30 10093.A1 10093.A7
10093.30EA 63.5 EUR
10093.30 10093.A1 10093.A7
Aluminium ≥99.9% (metals basis), granules 8-12 mm (0.32-0.47 in)
Aluminium
Purity: 99.9% CAS No.: 7429-90-5 Odour: Odourless MDL No.: MFCD00134029
Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321