Aluminium ≥99.999% (metals basis), rods, Puratronic®, Ø 22 mm (0.87 in)
Supplier: Thermo Fisher Scientific
Synonyms: Aluminum
10778.BQ 10778.BU
10778.BQEA 194 EUR
10778.BQ 10778.BU
Aluminium ≥99.999% (metals basis), rods, Puratronic®, Ø 22 mm (0.87 in)
Aluminium
Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈10.35g/cm Odour: Odourless MDL No.: MFCD00134029
Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321