Aluminium ≥99.99% (metals basis), foil, annealed, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
Synonyms: Aluminum
40531.FI 40531.GH 40531.HP
40531.FIEA 77.7 EUR
40531.FI 40531.GH 40531.HP
Aluminium ≥99.99% (metals basis), foil, annealed, Thickness: 0.5 mm (0.02 in)
Aluminium
Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈3.4g/50x50mm, Temper: soft Odour: Odourless MDL No.: MFCD00134029
Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321