Aluminium ≥99% (metals basis), spheres, Ø 2.38 mm (0.094 in)
Supplier: Thermo Fisher Scientific
Synonyms: Aluminum
42029.KM 42029.KN
42029.KMEA 56.3 EUR
42029.KM 42029.KN
Aluminium ≥99% (metals basis), spheres, Ø 2.38 mm (0.094 in)
Aluminium
Purity: 99% CAS No.: 7429-90-5 Molecular Formula: Alloy 1100, Al:Cu; 99+:0.12 wt% MDL No.: MFCD00213972
Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321