Aluminium ≥99.997% (metals basis), foil, Puratronic®, Thickness: 0.2 mm (0.008 in)
Supplier: Thermo Fisher Scientific
Synonyms: Aluminum
43776.GH 43776.HP
43776.GHEA 139 EUR
43776.GH 43776.HP
Aluminium ≥99.997% (metals basis), foil, Puratronic®, Thickness: 0.2 mm (0.008 in)
Aluminium
Purity: 99.997% CAS No.: 7429-90-5 Physical Form: ≈5.4g/100x100mm, Temper: as rolled Odour: Odourless MDL No.: MFCD00134029
Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321