Aluminium 99,9998 Metals basis, wire, Puratronic®, annealed, Ø 2.0 mm (0.08 in)

Supplier: Thermo Fisher Scientific

Synonyms: Aluminum

44335.G1 44335.G5
44335.G1EA 157 EUR
44335.G1 44335.G5
Aluminium 99,9998 Metals basis, wire, Puratronic®, annealed, Ø 2.0 mm (0.08 in)
Aluminium

Purity: 99.9998% CAS No.: 7429-90-5 Physical Form: ≈8.4g/m, Temper: soft Odour: Odourless MDL No.: MFCD00134029

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR