Aluminium ≥99.7% (metals basis), soluble APS 11 µm
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Aluminum
45031.A1 45031.A7 45031.30
45031.A1EA 217 EUR
45031.A1 45031.A7 45031.30
Aluminium ≥99.7% (metals basis), soluble APS 11 µm
Aluminium
Purity: 99.7% CAS No.: 7429-90-5 Sensitivity: Moisture Sensitive Appearance: Silver-grey Odour: Odourless UN No.: 3208 MDL No.: MFCD00134029
Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
UN: 3208
ADR: 4.3 II
Merck Index: 13,00321