Indium ≥99.999% (metals basis), wire, Puratronic®, Ø 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
00470.G1 00470.G5
00470.G1EA 152 EUR
00470.G1 00470.G5
Indium ≥99.999% (metals basis), wire, Puratronic®, Ø 1.0 mm (0.04 in)
Indium
Purity: 99.999% CAS No.: 7440-74-6 Physical Form: ≈5.74g/m Odour: Odourless MDL No.: MFCD00134048
Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971