Indium ≥99.9995% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)
Supplier: Thermo Fisher Scientific
10613.09 10613.18
10613.09EA 148 EUR
10613.09 10613.18
Indium ≥99.9995% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)
Indium
Purity: 99.9995% CAS No.: 7440-74-6 Odour: Odourless MDL No.: MFCD00134048
Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971