Indium ≥99.9999% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)
Supplier: Thermo Fisher Scientific
10614.09 10614.18
10614.09EA 160 EUR
10614.09 10614.18
Indium ≥99.9999% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)
Indium
Purity: 99.9999+% CAS No.: 7440-74-6 Odour: Odourless MDL No.: MFCD00134048
Formula: In
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971