Charcoal activated, Sigma-Aldrich®
Supplier: Merck
Synonyms: Carbon activated, Activated carbon, Activated charcoal
M-Clarity™ quality level = MQ200
C9157-2.5KG C9157-500G
SIALC9157-2.5KGEA 416 EUR
SIALC9157-2.5KG SIALC9157-500G
Charcoal activated, Sigma-Aldrich®
Charcoal activated
Formula: C
MW: 12.01 g/mol
Melting Pt: 3550
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2 III
REACH: 01-2119488894-16
Merck Index: 13,01818