Glassy carbon, powder 0,4-12 µm Type 1
Supplier: Thermo Fisher Scientific
Warning
38001.09 38001.18 38001.30
38001.09EA 54 EUR
38001.09 38001.18 38001.30
Glassy carbon, powder 0,4-12 µm Type 1
Glassy carbon
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1325
ADR: 4.1 III
Merck Index: 13,01818