Glassy carbon, spherical powder 0,4-12 µm Type 1
Supplier: Thermo Fisher Scientific
Warning
38004.09 38004.18 38004.30
38004.09EA 82.5 EUR
38004.09 38004.18 38004.30
Glassy carbon, spherical powder 0,4-12 µm Type 1
Glassy carbon
Formula: C
Melting Pt: 3652 °C
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1325
ADR: 4.1,III
Merck Index: 13,01818