Glassy carbon, rods, Ø 7 mm (0.28 in) Type 1
Supplier: Thermo Fisher Scientific
38012.DJ 38012.DM 38012.DR
38012.DJEA 332 EUR
38012.DJ 38012.DM 38012.DR
Glassy carbon, rods, Ø 7 mm (0.28 in) Type 1
Glassy carbon
Physical Form: ≈0.59g/cm MDL No.: MFCD00133992
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818