Glassy carbon, plate, Thickness: 1 mm (0.04 in) Type2
Supplier: Thermo Fisher Scientific
38021.FI 38021.GH
38021.FIEA 492 EUR
38021.FI 38021.GH
Glassy carbon, plate, Thickness: 1 mm (0.04 in) Type2
Glassy carbon
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818