Glassy carbon, plate, Thickness: 4 mm (0.16 in) Type 1
Supplier: Thermo Fisher Scientific
38026.FI 38026.GH 38026.FF
38026.FIEA 475 EUR
38026.FI 38026.GH 38026.FF
Glassy carbon, plate, Thickness: 4 mm (0.16 in) Type 1
Glassy carbon
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818