Glassy carbon, plate, Thickness: 3 mm (0.1 in) Type 2
Supplier: Thermo Fisher Scientific
42820.FF 42820.FI
42820.FFEA 152 EUR
42820.FF 42820.FI
Glassy carbon, plate, Thickness: 3 mm (0.1 in) Type 2
Glassy carbon
Formula: C
Melting Pt: 3652 °C
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818