Silicon, Sigma-Aldrich®
Supplier: Merck
MQ100
646687-1EA 646687-5EA
SIAL646687-1EAEA 203 EUR
SIAL646687-1EA SIAL646687-5EA
Silicon, Sigma-Aldrich®
Silicon
Formula: Si
MW: 28.09 g/mol
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635