Silicon, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
647535-1EA 647535-5EA
SIAL647535-1EAEA 360 EUR
SIAL647535-1EA SIAL647535-5EA
Silicon, Sigma-Aldrich®
Silicon
Formula: Si
MW: 28.09 g/mol
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635