Silicon, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
647543-1EA
647543-5EA
SIAL647543-1EAEA
258
EUR
SIAL647543-1EA
SIAL647543-5EA
Silicon, Sigma-Aldrich®
Silicon
Formula:
Si Boiling Pt: 2355 °C (1013 hPa) Melting Pt: 1410 °C Density: 2.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS Number: 7440-21-3 EINECS: 231-130-8 Merck Index: 12,08635 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...