Tin ≥99.9% (metals basis), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Fisher Scientific
14148.BU 14148.BY
14148.BUEA 134 EUR
14148.BU 14148.BY
Tin ≥99.9% (metals basis), rods, Ø 12.7 mm (0.5 in)
Tin
Purity: 99.9% CAS No.: 7440-31-5 Physical Form: ≈9.23g/cm Odour: Odourless MDL No.: MFCD00133862
Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587