Tin ≥99% (metals basis), foil, Thickness: 0.33 mm (0.013 in)
Supplier: Thermo Fisher Scientific
14636.RD 14636.RE
14636.RDEA 148 EUR
14636.RD 14636.RE
Tin ≥99% (metals basis), foil, Thickness: 0.33 mm (0.013 in)
Tin
Purity: 99+% CAS No.: 7440-31-5 Physical Form: ≈100g/15x27cm Odour: Odourless MDL No.: MFCD00133862
Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587